missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Polypropylene Plates

Manufactured from polypropylene for optimum visibility and high chemical resistance. Corning™ Polypropylene Plates are an excellent choice for transport and long-term storage.

Brand:  Corning™ 3657

387.90 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 10584391

  • £514.00 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or nonsterile

Order Info

Seal with Storage Mat III (sold separately).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
16 x 24
Polypropylene
Non-sterile
Round
100/Cs.
100/Cs.
Without Lid
Clear
384
Non-treated
Transport and longterm storage
25/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates
View accessory products
Special Offers

Special Offers

  • Multiple Manufacturers
    Save up to 94% on Teaching Lab Essentials
    Promo Code25721

    Expires2025-05-01

    New Offer
    Save Now
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Days

    6

    Hours

    22

    Minutes

    22

    Seconds

    23
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Polypropylene Plates > Clear; Standard; 95 μL; No; Round

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.