missing translation for 'onlineSavingsMsg'
Learn More
Corning™ Costar™ Disposable Traditional Style Polystyrene Storage Bottles Product Code.: 10177500
Add to basket

Corning™ Costar™ Disposable Traditional Style Polystyrene Storage Bottles
GREENER_CHOICEBIOPHARMA

Product Code. 10177500 Shop All Corning Products
1 L
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L

Product Code. 10177500

Brand: Corning™ 8396

Edge
Add to basket
Add to basket

Manufactured from clear polystyrene for strength and optimum visibility. Corning™ Costar™ Disposable Storage Bottles offer the ideal solution for storing media, buffers, and other aqueous solutions

  • Disposable polystyrene bottles for storage of media, buffers, and other aqueous solutions
  • Costar™ traditional style has smooth sides
  • Plug seal caps (45 mm) provide an airtight seal and help minimize the risk of contamination.
  • Bottles can be used with Corning vacuum filtration systems with 45 mm neck sizes (Cat. Nos. 430626, 430627, 431161, 430049, 430513, 430514,431118, 430015, and 431174).
  • Sterile
  • Nonpyrogenic
Greener Choice Claims
  • Environmental benefits include:
    • Energy Star Certification
    • Renewable Energy
    • Certifications and Seals of Approval
TRUSTED_SUSTAINABILITY

Specifications

Capacity 32 oz., 1 L
Capacity (Metric) 1 L
Capacity (English) 32 oz.
Color Clear
Material Polystyrene
No. per Case 12/Cs.
No. per Pack 1/Pk.
Sterility Sterile
Closure Color Red
Neck Finish 45
Shape Cylindrical
Closure Material HDPE
Closure Size 45 mm
Closure Type Plug Seal
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number30424006 Certificate TypeCertificate of Quality Date11/04/2025 Catalog Number8396
Lot Number01525001 Certificate TypeCertificate of Quality Date11/04/2025 Catalog Number8396
Lot Number07624003 Certificate TypeCertificate of Quality Date07/12/2024 Catalog Number8396
Lot Number23324005 Certificate TypeCertificate of Quality Date27/11/2024 Catalog Number8396
Lot Number07024002 Certificate TypeCertificate of Quality Date07/11/2024 Catalog Number8396
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Costar™ Disposable Traditional Style Polystyrene Storage Bottles >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.