missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CTS™ CytoTune™-iPS 2.1 Sendai Reprogramming Kit

The first off-the-shelf reprogramming system designed for clinical and translational research

Brand:  Invitrogen™ A34546

Product Code. 15853374

  • £13590.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The CTS™ CytoTune™-iPS 2.1 Sendai Reprogramming Kit is the first off-the-shelf reprogramming system designed for clinical and translational research. Like the CytoTune-iPS 2.0 Sendai reprogramming kit, this kit uses Sendai particles to deliver Yamanaka factors that are critical for efficient generation of induced pluripotent stem cells (iPSCs). This kit includes three vector preparations: polycistronic Klf4–Oct3/4–Sox2 (KOS), L-Myc, and Klf4.

Manufactured in accordance with GMP requirements:

This product is manufactured at a site with an ISO9001-certified quality management system. The facility and process were audited for conformity with current Good Manufacturing Practices for medical devices, 21 CFR Part 820 of the regulation.

Formulated animal origin–free:

The bovine serum albumin carrier protein was removed from the CTS CytoTune-iPS 2.1 Sendai vector formulation. The preparation contains no animal-derived components at the primary component level.

Accompanied by documentation to support your regulatory filing:

Full traceability documentation such as Certificate of Origin and Certificate of Analysis are available.

What you get:

The kit includes 0.2 mL of each vector (KOS, Klf4, and L-myc) at a titer of 0.5–2.0 x 108 CFU/mL, sufficient to reprogram a minimum of six wells of a six-well plate.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 0.2 mL CTS CytoTune KOS
• 0.2 mL CTS CytoTune Klf4
• 0.2 mL CTS CytoTune L-myc

Store at -70°C.
Cell and Gene Therapy Research, Development, and Manufacturing
iPSC Reprogramming Kit
Dry Ice
Sendai
CTS™
1 kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2LG190008 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA34546
Lot Number2LG200010 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA34546
Lot Number2LG220011 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberA34546
Lot NumberLG180007 Certificate TypeCertificate of Analysis Date29/09/2020 Catalog NumberA34546
Lot Number2LG180007 Certificate TypeCertificate of Analysis Date28/09/2020 Catalog NumberA34546
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ CTS™ CytoTune™-iPS 2.1 Sendai Reprogramming Kit > KitContents: 0.2 mL CTS CytoTune KOS; 0.2 mL CTS CytoTune Klf4; 0.2 mL CTS CytoTune L-myc

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.