Learn More
Ambion™ Custom Select siRNA
Custom Select siRNA
Brand: Ambion™ 4390830
This item is not returnable.
View return policy
Description
Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.
Specifications
Room temperature | |
Knocking down gene expression in order to study protein function in a wide range of cell types | |
HPLC |
Custom Select siRNA, 20nmol, HPLC Purity, ≥95% Purity, Ideal for Knocking Down Gene Expression in Order to Study Protein Function in a Wide Range of Cell Types, These siRNAs Provide High Knock-down Efficiencies and Specificities | |
Custom Select siRNA | |
20 nmol |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Product Content Correction
Product Title
Ambion™ Custom Select siRNA > 20 nmol