missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-Fructose, 99% Product Code.: 11463308

Thermo Scientific Chemicals D-Fructose, 99%

Product Code. 11463308
5000 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11463308

Brand: Thermo Scientific Chemicals A17718.0I

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fructose is used commercially in foods and beverages. It increases starch viscosity more rapidly and achieves a higher final viscosity than sucrose. It has all the fiber squeezed out of it and so the fructose is easily absorbed into the bloodstream. High-fructose-corn syrup is used in frozen products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fructose is used commercially in foods and beverages. It increases starch viscosity more rapidly and achieves a higher final viscosity than sucrose. It has all the fiber squeezed out of it and so the fructose is easily absorbed into the bloodstream. High-fructose-corn syrup is used in frozen products.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00148910
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
Synonym d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300Show MoreShow Less
PubChem CID 5984
ChEBI CHEBI:48095
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point ∼105°C to 110°C
Density 1.59
Assay Percent Range 99%
Odor Odorless
Quantity 5000 g
Beilstein 1239004
Merck Index 14,4273
Solubility Information Soluble in water.
Optical Rotation −89° (c=2 in Water)
Formula Weight 180.16
Percent Purity 99%
Chemical Name or Material D-Fructose
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252859 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog NumberA17718.30,A17718.0I,A17718.0B
Lot Number10251789 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog NumberA17718.30,A17718.0I,A17718.0B
Lot NumberM03G012 Certificate TypeCertificate of Analysis Date22/11/2022 Catalog NumberA17718.30,A17718.0I,A17718.0B
Lot Number10239052 Certificate TypeCertificate of Analysis Date15/11/2022 Catalog NumberA17718.30,A17718.0I,A17718.0B
Lot NumberT15I025 Certificate TypeCertificate of Analysis Date02/08/2022 Catalog NumberA17718.30,A17718.0I,A17718.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-Fructose, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.