missing translation for 'onlineSavingsMsg'
Learn More
D-(+)-Trehalose, Dihydrate, Fisher BioReagents Product Code.: 11413104

D-(+)-Trehalose, Dihydrate, Fisher BioReagents

Product Code. 11413104
25 g, Poly Bottle
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 11413104

Brand: Fisher BioReagents BP268725

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
D-(+)-Trehalose is used in several biochemical techniques.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6138-23-4
Molecular Formula C12H26O13
Molecular Weight (g/mol) 378.327
MDL Number MFCD00071594
InChI Key DPVHGFAJLZWDOC-PVXXTIHASA-N
Synonym trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triolShow MoreShow Less
PubChem CID 181978
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol;dihydrate
SMILES C(C1C(C(C(C(O1)OC2C(C(C(C(O2)CO)O)O)O)O)O)O)O.O.O

Specifications

Melting Point 97°C
Color White
Loss on Drying 8.5 to 10.5%
Infrared Spectrum Conforms to reference
Assay Percent Range ≥98 %
Packaging Poly Bottle
Quantity 25 g
Merck Index 15, 9742
Formula Weight 378.33
Percent Purity 98% min.
Physical Form Powder or Solid
Chemical Name or Material D-(+)-Trehalose, Dihydrate
Show More Show Less
Product Identifier
  • D(+)-Trehalose dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-(+)-Trehalose, Dihydrate, Fisher BioReagents >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.