missing translation for 'onlineSavingsMsg'
Learn More
Di-tert-butyl dicarbonate, 99%, Thermo Scientific Chemicals Product Code.: 10276630

Di-tert-butyl dicarbonate, 99%, Thermo Scientific Chemicals

Product Code. 10276630
100 g, Glass bottle
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10276630

Brand: Thermo Scientific Acros 194671000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24424-99-5
Molecular Formula C10H18O5
Molecular Weight (g/mol) 218.25
MDL Number MFCD00008805
InChI Key DYHSDKLCOJIUFX-UHFFFAOYSA-N
Synonym di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonateShow MoreShow Less
PubChem CID 90495
ChEBI CHEBI:48500
IUPAC Name tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
SMILES CC(C)(C)OC(=O)OC(=O)OC(C)(C)C

Specifications

Melting Point 22.0°C to 24.0°C
Color White
Density 1.0200g/mL
Boiling Point 56.0°C to 57.0°C (0.5 mmHg)
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula O[CO2C(CH3)3]2
Refractive Index 1.4075 to 1.4095
Quantity 100 g
Fieser 04,128; 07,91; 08,145; 10,122; 12,159; 13,94; 15,113
Merck Index 15, 3038
Specific Gravity 1.02
Formula Weight 218.25
Percent Purity 99%
Physical Form Low Melting Crystalline Solid
Chemical Name or Material Di-tert-butyl dicarbonate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462433 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number194670500,194671000,194670250,194675000,194670100
Lot NumberA0469151 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number194670500,194671000,194670250,194675000,194670100
Lot NumberA0458201 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number194670500,194671000,194670250,194675000,194670100
Lot NumberA0462433 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number194670500,194671000,194670250,194670100,194675000
Lot NumberA0443940 Certificate TypeCertificate of Analysis Date21/12/2023 Catalog Number194670500,194671000,194670250,194670100,194675000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Di-tert-butyl dicarbonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Di-tert-butyl dicarbonate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.