missing translation for 'onlineSavingsMsg'
Learn More
Diethyldithiocarbamic acid, sodium salt trihydrate, ACS reagent, Thermo Scientific Chemicals Product Code.: 10289370

Diethyldithiocarbamic acid, sodium salt trihydrate, ACS reagent, Thermo Scientific Chemicals

Product Code. 10289370
500 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10289370

Brand: Thermo Scientific Acros 424315000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20624-25-3
Molecular Formula C5H10NNaS2·3H2O
Molecular Weight (g/mol) 225.31
MDL Number MFCD00150617
InChI Key WWGXHTXOZKVJDN-UHFFFAOYSA-M
Synonym sodium diethyldithiocarbamate trihydrate, dithiocarb trihydrate, cupral, diethyldithiocarbamic acid sodium salt trihydrate, ditiocarb sodium trihydrate, unii-04574exf2c, diethyldithiocarbamate sodium trihydrate, carbamic acid, diethyldithio-, sodium salt, trihydrate, diethyldithiocarbamate sodium salt trihydrate, diethyldithiokarbaman sodny trihydrat czechShow MoreShow Less
PubChem CID 517546
IUPAC Name sodium;N,N-diethylcarbamodithioate;trihydrate
SMILES CCN(CC)C(=S)[S-].O.O.O.[Na+]

Specifications

Melting Point 95.0°C to 98.5°C
Color Gray to White
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (C2H5)2NCS2Na·3H2O
Quantity 500 g
Beilstein 04, II, 613
Sensitivity (to copper) Passes Test
Merck Index 15, 3421
Solubility Information (in water) Passes test
Formula Weight 225.31
Grade ACS Reagent
Physical Form Crystals or Powder
Chemical Name or Material Diethyldithiocarbamic acid, sodium salt trihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457402 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number424311000,424315000
Lot NumberA0444923 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number424311000,424315000
Lot NumberA0398146 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number424311000,424315000
Lot NumberA0406784 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number424311000,424315000
Lot NumberA0416070 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number424311000,424315000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyldithiocarbamic acid, sodium salt trihydrate, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.