missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Dispensette™ S Trace Analysis Analog-adjustable Bottletop Dispensers

The Dispensette S Trace Analysis bottle dispenser is for precise volume dispensing of high-purity media for trace analysis.The components of fluid path have been selected to only contain highest purity materials, such as fluoroplastics and sapphire. The result is trace metal content of dispensed liquid generally in the low ppb range, or even in low ppt range, depending on application.

Brand:  BRAND™ 4640040

732.70 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15408103

  • £896.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: Discharge tube, valve mounting/calibration tool, ETFE and PTFE adapters (GL28, GL32, S40), telescoping filling tube, recirculation tube (optional), performance certificate, operating manual
This item is not returnable. View return policy

Description

Description

  • Simple to mount discharge tube available with or without integrated recirculation valve
  • Designed without additional sealing rings for ease of cleaning and maintenance
  • Replaceable discharge valve with safety valve
  • Olive-shaped filling valve for firmer attachment of filling tubes
  • Improved flow technology results in faster priming
  • Scalloped track for secure volume setting on analog dispensers

Services

One year

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bottletop Dispenser
Without recirculation valve
1 to 10 mL
Dispensing acids, bases and hydrogen peroxide in the volume range from 1 to 10mL
ISO 9001
Platinum Iridium Valve Spring
Analog
Product Suggestions

Product Suggestions

Videos

Videos

  • Brand Dispensette S Accessories

  • Brand Dispensette S Cleaning

  • Brand Dispensette S Startup

  • Brand Dispensette S Teaser

SDS
Documents Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Dispensette™ S Trace Analysis Analog-adjustable Bottletop Dispensers > Without recirculation valve; Valve spring: Platinum-iridium

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.