Learn More
Interscience™ Dispensing Assembly for Dilutors
Set includes parts required for dispensing with dilutors.
Brand:  Interscience™ 513043
Product Code. 16322236
  • £256.88 / Each
Estimated Shipment: 24-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Includes:
  • Set of silicone tubings with inner diameter of 6.4 mm
  • GL45 screw cap
  • Tube weight
  • Nozzle
  • Filter
Specifications
Specifications
Dispensing Tubing
Autoclavable
Adding Dispensing Assembly to Dilutor
DiluFlow, DiluFlow Pro, DiluFlow Elite
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers
Product Content Correction
Product Title
Interscience™ Dispensing Assembly for Dilutors > Dispensing Assembly for GL45 (6.4 mm); For Use With (Equipment): DiluFlow, DiluFlow Pro, DiluFlow Elite