Learn More
Swann-Morton™ Disposable Scalpel Blade
Non-sterile, foil-packed, disposable scalpel blade. Swann-Morton™ Disposable Scalpel Blade is constructed of carbon steel.
Brand: Swann-Morton™ 0113
This item is not returnable.
View return policy
Description
- Non-sterile blade
- Foil packets of 5
- Fitment No.4. Fits handles 4, 4L, 4 Graduated and 6B
- Carbon steel construction
Specifications
Carbon Steel | |
Fitment # 4. Fits handles # 4, # 4L, # 4 Graduated & # 6B | |
Non-sterile | |
20 x 5/Pk. |
Scalpel Blade | |
Single-use | |
BS EN 7740 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Swann-Morton™ Disposable Scalpel Blade > Quantity: 20 x 5/Pk.
Spot an opportunity for improvement?Share a Content Correction