Learn More
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Round Bottom Flasks with Conical Ground Joint
Brand:  Lenz Laborglasinstrumente™ 03001928
 View more versions of this product
Product Code. 10697281
  • £91.90 / Pack of 10
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
According to DIN EN ISO 4797 and in addition to standard
Borosilicate Glass, Duran Glass
Round
50 mL
19/26 NS Socket
51 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Round Bottom Flasks with Conical Ground Joint > Capacity/Socket Joint: 50mL-19/26 NS Socket