Learn More
Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks
Manufactured from Duran™ glass. Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks include a conical ground joint.
£13.15 - £363.67
Specifications
Certifications/Compliance DIN EN ISO 4797
Material Borosilicate Glass, Duran Glass
Neck Style Medium
Bottom Shape Round
View More Specs
Products 8
Product Code Brand Capacity (Metric) Diameter (Metric) Outer Standard Taper Joint Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric) Outer Standard Taper Joint Price Quantity & Availability  
10483861
View Documents
Lenz Laborglasinstrumente™
03002970
1000 mL 131 mm NS 29/32
£210.00
Pack of 10
In Stock
to see stock.
Add to basket
15360895
View Documents
Lenz Laborglasinstrumente™
03001413
10 mL 31 mm NS 14/23
£110.42
Pack of 10
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
11737425
View Documents
Lenz Laborglasinstrumente™
03002937
100 mL 64 mm NS 29/32
£90.01
Pack of 10
In Stock
to see stock.
Edge
Add to basket
10211551
View Documents
Lenz Laborglasinstrumente™
03002949
250 mL 85 mm NS 29/32
£204.97
Pack of 10
In Stock
to see stock.
Add to basket
15370885
View Documents
Lenz Laborglasinstrumente™
03001422
25 mL 45 mm NS 14/23
£47.50
Pack of 10
In Stock
to see stock.
Add to basket
11915956
View Documents
Lenz Laborglasinstrumente™
03002958
500 mL 105 mm NS 29/32
£363.67
Pack of 10
In Stock
to see stock.
Add to basket
10710992
View Documents
Lenz Laborglasinstrumente™
03001428
50 mL 51 mm NS 14/23
£79.82
Pack of 10
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
10508851
View Documents
Lenz Laborglasinstrumente™
03002928
50 mL 51 mm NS 29/32
£13.15
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
Description
Description
  • Manufactured from Duran™ glass
  • Complies with DIN EN ISO 4797 standards
  • Includes conical ground joint
Specifications
Specifications
DIN EN ISO 4797
Borosilicate Glass, Duran Glass
Medium
Round
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks