missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks
Manufactured from Duran™ glass. Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks include a conical ground joint.
Brand: Lenz Laborglasinstrumente™ 03002970
Description
- Manufactured from Duran™ glass
- Complies with DIN EN ISO 4797 standards
- Includes conical ground joint
Specifications
DIN EN ISO 4797 | |
Borosilicate Glass, Duran Glass | |
NS 29/32 | |
131 mm |
1000 mL | |
Medium | |
Round |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks > Capacity: 1000mL; Frosted NS 29/32
Spot an opportunity for improvement?Share a Content Correction