missing translation for 'onlineSavingsMsg'
Learn More
(Ethoxycarbonylmethyl)dimethylsulfonium bromide, 98%, Thermo Scientific Chemicals Product Code.: 11309973

(Ethoxycarbonylmethyl)dimethylsulfonium bromide, 98%, Thermo Scientific Chemicals

Product Code. 11309973
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11309973

Brand: Thermo Scientific Alfa Aesar L00266.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5187-82-6
Molecular Formula C6H13BrO2S
Molecular Weight (g/mol) 229.132
MDL Number MFCD00830557
InChI Key JXFPTJYKYKVENJ-UHFFFAOYSA-M
Synonym ethoxycarbonylmethyl dimethylsulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulfonium bromide, carbethoxymethyl-dimethylsulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulfanium bromide, 2-ethoxy-2-oxoethyl dimethyl sulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulphonium bromide, sulfonium, carbethoxymethyl dimethyl-, bromide, sulfonium, 2-ethoxy-2-oxoethyl dimethyl-, bromide, carbethoxymethyl dimethylsulfonium bromide, acmc-1az78Show MoreShow Less
PubChem CID 78861
IUPAC Name (2-ethoxy-2-oxoethyl)-dimethylsulfanium;bromide
SMILES CCOC(=O)C[S+](C)C.[Br-]

Specifications

Melting Point ∽80°C (decomposition)
Quantity 50 g
Beilstein 4161472
Sensitivity Hygroscopic
Formula Weight 229.14
Percent Purity 98%
Chemical Name or Material (Ethoxycarbonylmethyl)dimethylsulfonium bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250878 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL00266.18,L00266.09
Lot Number10250063 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog NumberL00266.18,L00266.09
Lot Number10170170 Certificate TypeCertificate of Analysis Date04/07/2023 Catalog NumberL00266.18,L00266.09,L00266
Lot Number10170170 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL00266.18,L00266.09
Lot Number10142967 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL00266.18,L00266.09
compliance-icons
Product Identifier
  • (Ethoxycarbonylmethyl)dimethylsulfoniumbromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : (2-Ethoxy-2-oxoethyl)dimethylsulphoniumbromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(Ethoxycarbonylmethyl)dimethylsulfonium bromide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.