Learn More
Heidolph™ Evaporating Flask
For use with rotary evaporators
Brand:  Heidolph™ 514-70000-00
 View more versions of this product
Product Code. 10251292
  • £110.00 / Each
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Evaporating flasks come with a standard NS 29/32 joint size but are also available in NS 24/29
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Evaporating Flask
50 mL
Glass
Rotary evaporators
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Heidolph™ Evaporating Flask > Evaporating flask 50 mL