Learn More
Falcon™ Tissue Culture Treated Flasks
Manufactured from optically clear virgin polystyrene for easy viewing of sample or cells. Falcon™ Tissue Culture Treated Flasks features vacuum-gas plasma tissue culture treatment to ensure consistent growth surface.
£102.92 - £294.87
Specifications
Closure Material | Polyethylene |
---|---|
Material | Polystyrene |
For Use With (Application) | For enhanced cell attachment and growth |
Shape | Rectangular |
Surface Coating | TC-Treated |
Product Code | Brand | Capacity (Metric) | Culture Area | Closure Type | Neck Style | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Culture Area | Closure Type | Neck Style | Price | Quantity & Availability | |||
10354331
![]() ![]() |
Falcon™
353018 |
25 mL | 12.5 cm2 | Plug Seal | Canted |
£174.50
Pack of 100 |
In Stock
Log in to see stock. |
|||
10249120
![]() ![]() |
Falcon™
353107 |
25 mL | 12.5 cm2 | Vented | Canted |
£194.49
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||
10634501
![]() ![]() |
Falcon™
353014 |
50 mL | 25 cm2 | Plug Seal | Canted |
£237.31
Pack of 200 |
In Stock
Log in to see stock. ![]() |
|||
10350391
![]() ![]() |
Falcon™
353108 |
50 mL | 25 cm² | Vented | Canted |
£102.92
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||
10341011
![]() ![]() |
Falcon™
353082 |
70 mL | 25 cm2 | Plug Seal | Canted |
£244.43
Pack of 200 |
In Stock
Log in to see stock. |
|||
10700052
![]() ![]() |
Falcon™
353109 |
70 mL | 25 cm2 | Vented | Canted |
£153.33
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||
10527161
![]() ![]() |
Falcon™
353135 |
250 mL | 75 cm² | Plug Seal | Canted |
£150.37
Pack of 60 |
In Stock
Log in to see stock. |
|||
10797791
![]() ![]() |
Falcon™
353024 |
250 mL | 75 cm2 | Plug Seal | Straight |
£254.36
Pack of 100 |
Estimated Shipment: 22-05-2025
Log in to see stock. |
|||
10537161
![]() ![]() |
Falcon™
353136 |
250 mL | 75 cm2 | Vented | Canted |
£170.92
Pack of 60 |
In Stock
Log in to see stock. ![]() |
|||
10508662
![]() ![]() |
Falcon™
353110 |
250 mL | 75 cm2 | Vented | Straight |
£294.87
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||
10728502
![]() ![]() |
Falcon™
355000 |
600 mL | 150 cm2 | Plug Seal | Canted |
£171.24
Pack of 40 |
In Stock
Log in to see stock. |
|||
10499032
![]() ![]() |
Falcon™
355001 |
600 mL | 150 cm2 | Vented | Canted |
£193.16
Pack of 40 |
In Stock
Log in to see stock. |
|||
10078780
![]() |
Falcon™
353028 |
750 mL | 175 cm2 | Plug Seal | Straight |
£240.95
Pack of 40 |
In Stock
Log in to see stock. |
|||
10218550
![]() ![]() |
Falcon™
353112 |
750 mL | 175 cm2 | Vented | Straight |
£280.74
Pack of 40 |
In Stock
Log in to see stock. ![]() |
|||
10446421
![]() ![]() |
Falcon™
353139 |
800 mL | 225 cm2 | Plug Seal | Canted |
£252.79
Pack of 30 |
In Stock
Log in to see stock. |
|||
10687762
![]() ![]() |
Falcon™
353138 |
800 mL | 225 cm2 | Vented | Canted |
£285.02
Pack of 30 |
In Stock
Log in to see stock. ![]() |
|||
Description
- Secure stacking provides stability from accidental tips or spills
- Nonpyrogenic tested to less than 0.1 EU/ mL
- Volumetric graduations and writing patch
- Vented caps incorporate a 0.2 μm hydrophobic membrane for optimal gas exchange
- Growth area and volume are nominal
Specifications
Polyethylene | |
For enhanced cell attachment and growth | |
TC-Treated | |
Rectangular |
Polystyrene | |
Rectangular | |
Sterile |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.