missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Selective D1-like dopamine receptor partial agonist (EC50 = 57 nM). Vasodilator in vivo and does not readily cross the blood-brain barrier. Also α2-adrenoceptor antagonist in vitro (Ki = 15 - 25 nM).

Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 342.22 |
Percent Purity | >98% |
Chemical Name or Material | Fenoldopam hydrochloride |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fenoldopam hydrochloride, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction