Learn More
Cole-Parmer™ Lab Markers
Write on a variety of surfaces with these versatile lab markers. Cole-Parmer™ Lab Markers permanently mark plastic; wash off glass, metal, or porcelain. Markers won’t bleed or smudge when subjected to water or alcohol.
Brand:  Cole-Parmer™ 09964-20
 View more versions of this product
Product Code. 11710239
Description
Description
  • Fine-point fiber tip
  • Quick-drying ink
  • Writes on cold, wet surfaces
  • Available in four different colors
Specifications
Specifications
Red
For Glass, Porcelain or Metal surfaces
10/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Lab Markers > Red