missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Fluorescein Product Code.: 11327996

Thermo Scientific Chemicals Fluorescein

Product Code. 11327996
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11327996

Brand: Thermo Scientific Chemicals L13251.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorescein is used as a fluorescent tracer for many applications including in a type of dye laser as the gain medium, in forensics and serology to detect latent blood stains and in dye tracing. It is used to localise multiple muscular ventricular septal defects during open heart surgery and confirm the presence of any residual defects. It is applied to teeth to reveal plaque.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water and alcohol.

Notes
Incompatibles with strong oxidizing agents. Moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2321-07-5
Molecular Formula C20H12O5
Molecular Weight (g/mol) 332.31
MDL Number MFCD00005050
InChI Key GNBHRKFJIUUOQI-UHFFFAOYSA-N
Synonym fluorescein, solvent yellow 94, resorcinolphthalein, yellow fluorescein, 3,6-fluorandiol, d and c yellow no. 7, fluoresceine, japan yellow 201, c.i. solvent yellow 94, d&c yellow no. 7Show MoreShow Less
PubChem CID 16850
ChEBI CHEBI:31624
IUPAC Name 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O

Specifications

Melting Point 320°C
Quantity 100 g
Beilstein 94324
Merck Index 14,4159
Solubility Information Slightly soluble in water and alcohol.
Formula Weight 332.32
Percent Purity ≥90%
Chemical Name or Material Fluorescein

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249796 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog NumberL13251.22,L13251.36
Lot Number10248142 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog NumberL13251.22,L13251.36
Lot Number10239098 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL13251.22,L13251.36
Lot Number10172004 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL13251.22,L13251.36
Lot Number10211918 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL13251.22,L13251.36
compliance-icons
Product Identifier
  • Fluorescein
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Fluorescein

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Fluorescein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.