Learn More
Gilson™ Tip Empty Rack For Pipet Tips
GREENER_CHOICE
Use with loose Gilson Diamond tips. Gilson™ Tip Empty Rack For Pipette Tips are color coded for ease of use.
Brand:  Gilson™ F171402
 View more versions of this product
Product Code. 12619601
  • £19.10 / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Fully autoclavable
  • Made of polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Empty Rack
Blue/Green
Polypropylene
Yes, Interlocking
Diamond Tips
3.66 in.
4.88 in.
Autoclavable
Pipette Tips (20 to 300 μL)
Rectangular
Non-sterile
9.3 cm
5.28 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gilson™ Tip Empty Rack For Pipet Tips > Tip Empty Rack For Pipet Tips; Holds: 20 to 300 μL D300 Pipet Tips