Learn More
Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispenser
Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispensers have a dispensing cylinder and piston made of glass.
Brand:  Poulten Graf™ 101 08144
 View more versions of this product
Product Code. 10558721
  • £556.00 / Each
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For dispensing of pure organic solvents. Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispensers have a dispensing cylinder and piston made of glass.
  • Manufactured according to DIN EN ISO 9001:2008 using high quality materials
  • Solid dispensing piston made of glass
  • Cylinder coated with plastic to protect against breakage
  • With certificate, individually numbered
  • Ergonomic finger protection cap made of PP with volume setting system
  • Graduation on the scale is lasered and easy to read
  • Fast and easy volume setting with additional fine adjustment
Specifications
Specifications
Fortuna Optifix Solvent Dispenser
PTFE, Polypropylene, Fiberglass
For dispensing solvents
1
6 to 30 mL
Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Poulten Graf™ Glass/PP/PTFE Fortuna™ Optifix™ Solvent Dispenser > Volume: 6 to 30 mL