missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Glycine, 0.2M buffer soln., pH 2.5 Product Code.: 15454349

Thermo Scientific Chemicals Glycine, 0.2M buffer soln., pH 2.5

Product Code. 15454349
250 mL
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy

Product Code. 15454349

Brand: Thermo Scientific Chemicals J61855.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This buffer solution is used for eluting (dissociating) antibodies in various antibody purification procedures such as affinity purification of antibody using peptide coupled to a column. It is widely used as a buffer for a variety of immunological applications. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This buffer solution is used for eluting (dissociating) antibodies in various antibody purification procedures such as affinity purification of antibody using peptide coupled to a column. It is widely used as a buffer for a variety of immunological applications. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

Solubility
Soluble in water (100 mg/ml), and acetone (slightly soluble). Insoluble in alcohol, and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature 2 - 8°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Concentration 0.2 M
pH 2.5
Physical Form Liquid
Quantity 250 mL
Odor Odorless
Chemical Name or Material Glycine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP19L570 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog NumberJ61855.AP,J61855.AK
Lot NumberN25L501 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog NumberJ61855.AP,J61855.AK
Lot NumberM16L556 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberJ61855.AP,J61855.AK
Lot NumberY25K522 Certificate TypeCertificate of Analysis Date27/11/2024 Catalog NumberJ61855.AP,J61855.AK
Lot NumberX20K534 Certificate TypeCertificate of Analysis Date27/11/2024 Catalog NumberJ61855.AP,J61855.AK

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Glycine, 0.2M buffer soln., pH 2.5 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.