missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals HEPES, 1.0M buffer soln., pH 7.5 Product Code.: 15493429

Thermo Scientific Chemicals HEPES, 1.0M buffer soln., pH 7.5

Product Code. 15493429
250 mL
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy

Product Code. 15493429

Brand: Thermo Scientific Chemicals J60712.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HEPES is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.              
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Concentration 1.0 M
pH 7.5
Physical Form Liquid
Quantity 250 mL
Solubility Information Miscible with water.
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
Chemical Name or Material HEPES buffer soln.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP15L524 Certificate TypeCertificate of Analysis Date12/03/2025 Catalog NumberJ60712.AK,J60712.AP
Lot NumberM11L542 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberJ60712.AK,J60712.AP
Lot NumberY10K500 Certificate TypeCertificate of Analysis Date27/11/2024 Catalog NumberJ60712.AK,J60712.AP
Lot NumberX28K527 Certificate TypeCertificate of Analysis Date27/11/2024 Catalog NumberJ60712.AK,J60712.AP
Lot NumberU02K502 Certificate TypeCertificate of Analysis Date16/09/2024 Catalog NumberJ60712.AK,J60712.AP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals HEPES, 1.0M buffer soln., pH 7.5 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.