missing translation for 'onlineSavingsMsg'
Learn More
HEPES, 1M Solution, pH 7.3, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals Product Code.: 15624198

HEPES, 1M Solution, pH 7.3, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Product Code. 15624198 Shop All Thermo Scientific Products
100 mL, Nalgene bottle
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene bottle
This item is not returnable. View return policy

Product Code. 15624198

Brand: Thermo Scientific J16924.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Concentration 1 M
pH 7.3
Physical Form Solution
Quantity 100 mL
Packaging Nalgene bottle
Beilstein 883043
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Grade Molecular Biology
Chemical Name or Material HEPES

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number245631 Certificate TypeCertificate of Analysis Date12/12/2024 Catalog NumberJ16924.K2,J16924.AE,J16924.AP
Lot Number242238 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ16924.K2,J16924.AE,J16924.AP
Lot Number235918 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ16924.K2,J16924.AE,J16924.AP
Lot Number234375 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberJ16924.K2,J16924.AE,J16924.AP
Lot Number198695 Certificate TypeCertificate of Analysis Date15/11/2023 Catalog NumberJ16924.K2,J16924.AE,J16924.AP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
HEPES, 1M Solution, pH 7.3, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.