missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals HEPES, 99% Product Code.: 11422497

Thermo Scientific Chemicals HEPES, 99%

Product Code. 11422497
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11422497

Brand: Thermo Scientific Chemicals A14777.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HEPES is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.  
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym 4-(2-Hydroxyethyl)-1-piperazineethanesulfonic acid
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Melting Point ∼236°C (decomposition)
Quantity 250 g
Assay Percent Range 99%
Beilstein 883043
Merck Index 14,4654
Solubility Information Soluble in water.
Formula Weight 238.31
Percent Purity 99%
Chemical Name or Material HEPES

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250819 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14777.30,A14777.18,A14777.09
Lot Number10252179 Certificate TypeCertificate of Analysis Date23/08/2024 Catalog NumberA14777.30,A14777.18,A14777.09
Lot Number10248293 Certificate TypeCertificate of Analysis Date23/10/2023 Catalog NumberA14777.30,A14777.18,A14777.09
Lot Number10247764 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog NumberA14777.30,A14777.18,A14777.09
Lot Number10246859 Certificate TypeCertificate of Analysis Date09/06/2023 Catalog NumberA14777.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals HEPES, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.