missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Hibernate™-E Medium
Hibernate™-E Medium
Brand: Gibco™ A1247601
126.35 GBP valid until 2024-06-28
Use promo code "21689" to get your promotional price.
Code : 50
Additional Details : Weight : 0.64050kg
Description
Hibernate Medium is a CO2-independent nutrient medium for the maintenance of neural cells, tissue, and tissue slices. Hibernate-E Medium is optimized for embryonic cells or tissue in ambient carbon dioxide levels.Hibernate Medium enables neurons to be maintained in ambient CO2 levels for up to 48 hours. This provides scientists with greater convenience and control while running bench-top live cell experiments, such as microscopy, flow cytometry, and other physiology studies. Hibernate Medium also allows for the preservation of viable brain tissue before processing for up to a month with refrigeration and maintenance of tissue slices.
Hibernate Medium may be combined with B-27 Supplements to yield a complete medium.
For other applications, we provide alternative formulations of Hibernate Medium and B-27 Supplements.
For established protocols using Hibernate Medium, access our Gibco Neurobiology Protocol Handbook.
View publications using Gibco neurobiology products at our Gibco CellCite.
Specifications
Store in refrigerator (2–8°C) and protect from light. |
|
Serum-free | |
Liquid | |
Serum-free | |
Phenol Red | |
Ambient |
Neural Stem Cells | |
Mammalian Cell Culture | |
Stem Cell Media | |
Sterile | |
Hibernate™ | |
Cytotoxicity, Endotoxin, Sterility, pH, Osmolality |
For Research Use Only. Not for use in diagnostic procedures.