Learn More
Brannan™ Precision Lab Thermometer with 3-points Traceable Calibration
High precision lab thermometer complete with calibration certificate.
£94.20 - £147.00
Specifications
Certifications/Compliance | UK Traceable to National Standards ISO 9001 |
---|---|
Material | Glass |
Accuracy | ± 1.0°C |
Width (Metric) | 1.2 cm |
Product Code | Brand | Temperature Range (Metric) | Subdivision | Immersion Type | Length (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Subdivision | Immersion Type | Length (Metric) | Price | Quantity & Availability | ||
17863298
|
Brannan™
50/521/0 |
-20°C to +110°C | 0.5°C | Partial Immersion | 30.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17883018
|
Brannan™
50/504/0 |
-1°C to 101°C | 0.2°C | Partial Immersion | 40.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17873298
|
Brannan™
50/506/0 |
-1°C to 250°C | 1°C | Partial Immersion | 40.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17883038
|
Brannan™
50/552/0 |
-50°C to +50°C | 1°C | Partial Immersion | 30.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17803308
|
Brannan™
50/500/0 |
0°C to 40°C | 0.1°C | Partial Immersion | 40.5 cm |
£97.21
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17873038
|
Brannan™
50/531/0 |
-10°C to +150°C | 1°C | Partial Immersion | 20.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17822988
|
Brannan™
50/512/0 |
-1°C to 51°C | 0.1°C | Total Immersion | 40.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17813038
|
Brannan™
50/513/0 |
-1°C to 51°C | 0.2°C | Total Immersion | 40.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17803038
|
Brannan™
50/520/0 |
-35°C to +50°C | 0.5°C | Partial Immersion | 30.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17823018
|
Brannan™
50/516/0 |
-1°C to 250°C | 1°C | Total Immersion | 40.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17873018
|
Brannan™
50/501/0 |
0°C to 40°C | 0.2°C | Partial Immersion | 40.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17893028
|
Brannan™
50/511/0 |
0°C to 40°C | 0.2°C | Total Immersion | 40.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17823038
|
Brannan™
50/526/0 |
-20°C to +110°C | 0.5°C | Total Immersion | 30.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17833308
|
Brannan™
50/527/0 |
-20°C to +150°C | 0.5°C | Total Immersion | 30.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17853308
|
Brannan™
50/555/0 |
-100°C to +50°C | 1°C | Total Immersion | 30.5 cm |
£147.00
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17843308
|
Brannan™
50/505/0 |
-1°C to 201°C | 0.5°C | Partial Immersion | 40.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17853078
|
Brannan™
50/503/0 |
-1°C to 51°C | 0.2°C | Partial Immersion | 40.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17883068
|
Brannan™
50/514/0 |
-1°C to 101°C | 0.2°C | Total Immersion | 40.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17843118
|
Brannan™
50/536/0 |
-10°C to +150°C | 1°C | Total Immersion | 20.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17863048
|
Brannan™
50/510/0 |
0°C to 40°C | 0.1°C | Total Immersion | 40.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17813108
|
Brannan™
50/523/0 |
-10°C to +50°C | 0.5°C | Partial Immersion | 30.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17893298
|
Brannan™
50/525/0 |
-35°C to +50°C | 0.5°C | Total Immersion | 30.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17823308
|
Brannan™
50/522/0 |
-20°C to +150°C | 0.5°C | Partial Immersion | 30.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17833118
|
Brannan™
50/530/0 |
-10°C to +110°C | 1°C | Partial Immersion | 20.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17813308
|
Brannan™
50/515/0 |
-1°C to 201°C | 0.5°C | Total Immersion | 40.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17823118
|
Brannan™
50/502/0 |
-1°C to 51°C | 0.1°C | Partial Immersion | 40.5 cm |
£97.21
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17853118
|
Brannan™
50/535/0 |
-10°C to +110°C | 1°C | Total Immersion | 20.5 cm |
£94.20
Each |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- A range of spirit filled, high precision thermometers which are supplied with a calibration certificate
- Safe non-mercury filling
- Presented in a study cardboard cylinder for storage
- Made in UK
Specifications
UK Traceable to National Standards ISO 9001 | |
Glass | |
± 1.0°C | |
1.2 cm |