Learn More
Sperian™ Honeywell™ Matrix Uncorded Earplugs Product Code.: 11862741
Sperian™ Honeywell™ Matrix Uncorded Earplugs
Product Code. 11862741
Orange
Click to view available options
Color:
Blue
Orange
Product Code. 11862741
Brand: Sperian™ 1011236
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Offering enhanced communication, safety and compliance. Sperian™ Honeywell™ Matrix Uncorded Earplugs make insertion and removal easy with a patended no-roll design.
  • Available in either blue or orange
  • Uniform attenuation profile blocks loud noise while voice and other higher frequencies can be heard naturally
  • Smaller diameter eliminated that "plugged up" feeling in the ear canal
  • Smooth outer skin is a great match for the sensitive skin inside the ear canal
TRUSTED_SUSTAINABILITY
Specifications
Color Orange
Size Universal
Quantity 200/Pk.
Plug Shape Cylinder
Product Type Earplug
Disposable Single-use
Product Content Correction
Product Title
Sperian™ Honeywell™ Matrix Uncorded Earplugs >