Learn More
SCAT™ HPLC Supply and Waste Set, V2.0, Suitable for All HPLC Systems
The all-round carefree package suitable for all HPLC and UHPLC systems.
Brand: SCAT™ 307448
Description
- Protects your system from contamination
- Keeps solvent mixtures stable
- High performance materials with excellent chemical resistance
The safety set connects tubing safely and blocks hazardous vapours. Air valves and exhaust filters provide safe pressure equalisation without under- or overpressure in solvent bottles and waste containers
Specifications
Liquid Chromatography | |
4 x Safety Cap III, GL45, 8 x air valve, 1 x Safety Waste Cap, S60/61, electrostatic conductive, 1 x Exhaust filter L, V3.0, with splash guard and change indicator, service life 12 month, 1 x 5 litre canister, PE-HD-EL, 1 x Grounding cable, 2x clamp, L = 1,5 m, 4 x Laboratory bottle DURAN, GL45, 1000 ml, clear glass and round, with cap and pouring ring , 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 - 8 mm ID, 1 x connector, curved, 5.0 - 11.5mm ID, 1 x connector, angled, 9.5 - 10 mm ID | |
Complete Solvent Collection System |
Any HPLC System | |
4 | |
HPLC Supply and Disposal Set (12 Month), Electrostatic Conductive |
Product Suggestions
Customers who viewed this item also viewed