Learn More
Invitrogen™ NuPAGE™ MOPS SDS Running Buffer (20X)
Made with high-purity reagents and are strictly quality controlled.
Brand: Invitrogen™ NP000102
373.32 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Includes: NuPAGE MOPS SDS Running Buffer (20X)
Requires : The buffers are provided as a concentrated solution and require simple dilution with deionized water before use.
This item is not returnable. View return policy
Description
NuPAGE MOPS SDS Running Buffer (20X) is formulated for running proteins on NuPAGE Novex Bis-Tris gels only. NuPAGE™ MOPS SDS Running Buffer is recommended for separating small- to medium-sized proteins.
- Get consistent, convenient, high-quality results
- Pre-mixed NuPAGE buffers are convenient way to ensure high-quality, consistent electrophoresis results
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature

Specifications
Running Buffer | |
7.7 | |
NuPAGE | |
50 mM MOPS, 50 mM Tris Base, 0.1% SDS, 1 mM EDTA, pH 7.7, 0.01-0.09% N,N-dimethylformamide |
20X | |
Liquid | |
5 L |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3025185 | Certificate of Analysis | 31/01/2025 | NP000102,NP000102_LITL18 PROMO |
3059087 | Certificate of Analysis | 31/01/2025 | NP000102,NP000102_LITL18 PROMO |
3086061 | Certificate of Analysis | 31/01/2025 | NP000102,NP000102_LITL18 PROMO |
2476987 | Certificate of Analysis | 29/01/2025 | NP000102,NP000102_LITL18 PROMO |
2491779 | Certificate of Analysis | 29/01/2025 | NP000102,NP000102_LITL18 PROMO |
Special Offers
-
Multiple Manufacturers
New Offer Shop Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.