missing translation for 'onlineSavingsMsg'
Learn More
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals Product Code.: 15418995

Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15418995
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15418995

Brand: Thermo Scientific Alfa Aesar H35174.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 859833-15-1
Molecular Formula C8H6Cl2N2O
Molecular Weight (g/mol) 217.049
MDL Number MFCD07772809
InChI Key LHLAJCRPSZDOFF-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-6-carbonyl chloride hydrochloride, imidazo 1,2-a pyridine-6-carbonylchloride, hydrochloride 1:1, 4-hydroimidazo 1,2-a pyridine-6-carbonyl chloride, chloride, imidazo 1,2-a pyridine-6-carbonyl chloride-hydrogen chloride 1/1Show MoreShow Less
PubChem CID 18525720
IUPAC Name imidazo[1,2-a]pyridine-6-carbonyl chloride;hydrochloride
SMILES C1=CC2=NC=CN2C=C1C(=O)Cl.Cl

Specifications

Melting Point 248°C to 250°C
Quantity 250 mg
UN Number UN3261
Sensitivity Hygroscopic
Formula Weight 217.06
Percent Purity 95%
Chemical Name or Material Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10179508 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH35174.MD,H35174.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.