missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride , Tech., Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 80.0 |
CAS Max % | 100.0 |
Packaging | Amber Glass Bottle |
Linear Formula | Yes |
Quantity | 1 g |
Formula Weight | 217.05 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | Imidazo[1, 2-a]pyridine-6-carbonyl chloride hydrochloride |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride , Tech., Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction