Learn More
Thermo Scientific™ Immuno Tubes and Stoppers
Reproduce results time after time with these immuno tubes and stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays.
Brand: Thermo Scientific™ 466982
256.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Reproduce results time after time with Thermo Scientific™ Immuno Tubes and Stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays. These products are backed by industry-leading quality standards and certifications.
Features:- Offered in choice of three surfaces for optimal binding
- Three tube sizes with stoppers available (ordered separately)
- Fits standard equipment
Choice of Surface:
- Thermo Scientific™ PolySorp™ is a hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains.
- Thermo Scientific™ MaxiSorp™ is a hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character.
- Thermo Scientific™ Mini-Sorp™ is a polyethylene-based material that provides a low-binding surface.
Liquid Phase Immunoassay Tubes (Cat. Nos. 443990, 468608):
- MiniSorp surface
- Molded from special polyethylene formulation
- For liquid phase immunoasssays, including RIA, for storing of reagents, or collecting chromatographic fractions
Solid Phase Immunoassay Tubes:
- PolySorp and MaxiSorp surfaces available for choice of optimal binding
- For solid phase immuno techniques such as IRMA, ELISA and ILMA
- Certified binding homogeneity
- High–quality polystyrene for optimal quality
- StarTube design gives higher sensitivity and faster assay times
- Enhances assay sensitivity and reduces incubation times
Recommended for:
- ELISA
- Solid Phase RIA
- Liquid Phase RIA
- Sample storage

Specifications
Polyethylene | |
MiniSorp | |
11 mm | |
3600 |
Immuno Tubes and Stopper | |
70 mm | |
150 | |
4 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.