Learn More
Thermo Scientific™ Insight™ Pro Software
Streamline your routine, complex and advanced routines and workflows with Thermo Scientific™ Insight™ Pro Software. Prevent costly sampling and measurement errors with prompting, step-by-step workflows.
Brand:  Thermo Scientific™ 837-003900
 View more versions of this product
Product Code. 17389253
  • £1015.00 / Each
Explore more special offers
Description
Show More
Specifications
Specifications
Insight Pro Software
For use with Thermo Scientific Evolution One, One Plus, and Pro UV-Vis Spectrophotometers
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Insight™ Pro Software >