Learn More
Invitrogen™ Oligofectamine™ Transfection Reagent
Proprietary formulation for transfecting oligonucleotides and short interfering RNA (siRNA) into eukaryotic cells
Brand: Invitrogen™ 12252011
557.92 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
566.72 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Includes: Contains one vial (1mL) Oligofectamine Reagent.
Description
Oligofectamine Transfection Reagent forms stable complexes with oligos, permitting efficient transfection into eukaryotic cells in highly specific, yet nontoxic fashion. Oligofectamine Reagent is suitable for nuclear and cytoplasmic targets and transfects wide variety of cell lines including CHO, HEK-293, NIH 3T3, and HeLa.
Using Oligofectamine Transfection Reagent
- Oligofectamine Reagent is easy to use because it provides simple and fast protocol
- Just dilute Oligofectamine Reagent, mix with oligonucleotide, and add to your cells
- Oligofectamine Reagent requires nanomolar quantities of antisense oligonucleotide, reducing the amount of valuable oligonucleotide needed by up to thousand-fold
- Ideal for high-throughput applications
- Shown to work for siRNA transfections and is recommend when performing RNAi knockdown experiments in HeLa cells
DNA & RNA Purification & Analysis, RNA Extraction, RNAi, Epigenetics & Noncoding RNA Research, Total RNA Isolation, Total RNA from Animal Cells & Tissues, Total RNA from Bacterial Samples, Total RNA from Plant Cells, Total RNA from Yeast, miRNA & Noncoding RNA Analysis, miRNA Isolation
Order Info
Shipping Condition: Wet Ice

Specifications
Contains one vial (1 ml) Oligofectamine™ Reagent. Store at 4°C. Do not freeze. | |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Lipid-based Transfection | |
High-throughput Compatible | |
Transfection Reagent | |
No |
Established Cell Lines, Primary Cells, Hard-to-Transfect Cells | |
Synthetic siRNA | |
Transfection | |
Oligofectamine™ | |
1 mL | |
Wet Ice |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3056370 | Certificate of Analysis | 31/01/2025 | 12252011 |
2462041 | Certificate of Analysis | 29/01/2025 | 12252011 |
2498106 | Certificate of Analysis | 29/01/2025 | 12252011 |
CN2548383 | Certificate of Analysis | 29/01/2025 | 12252011 |
2546435 | Certificate of Analysis | 29/01/2025 | 12252011 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.