Learn More
Thermo Scientific™ KaryoStat+™ with Cell ID Assay Service
KaryoStat+ with Cell ID Assay Service is a fast and cost-effective alternative to typical G-banded karyotyping that evaluates both genetic stability and SNP-based cell identification.
Brand: Thermo Scientific™ A52986
This item is not returnable.
View return policy
Description
- A fast and cost-effective alternative to typical G-banded karyotyping that evaluates both genetic stability and SNP-based cell identification
- KaryoStat+ service leverages the array-based CytoScan HT-CMA 96F Assay Kit (906025), offering whole-genome coverage for accurate detection of chromosomal aberrations in human primary and stem cells
- Cell ID leverages the KaryoStat Assay (905403) to compare genotypes across samples
The KaryoStat+ with Cell ID Assay Service offers:
- Fast data delivery – turnaround time of four weeks or less
- Convenient sample prep – assay uses frozen cell pellet, eliminating need for shipping live cells
- Higher resolution – microarray resolution five times higher than traditional G-banding
- Exceptional support – a dedicated project manager for frequent project communication
- Broad cell type compatibility – all non-transformed human cell types accepted
Specifications
1 sample | |
Assay Service | |
Research Use Only | |
Human |
3 Weeks | |
Applied Biosystems™ | |
Stem Cell and Immuno-oncology Researchers |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Product Title
Thermo Scientific™ KaryoStat+™ with Cell ID Assay Service > 1 Sample
Spot an opportunity for improvement?Share a Content Correction