missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-alpha-Lecithin, granular, from soybean oil Product Code.: 10494501

Thermo Scientific Chemicals L-alpha-Lecithin, granular, from soybean oil

Product Code. 10494501
1 kg, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10494501

Brand: Thermo Scientific Chemicals 413100010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-alpha-Lecithin, >-97% (phospholipids) min. (as acetone insoluble), C42H80NO8P, CAS Number-8002-43-5

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8002-43-5
Molecular Formula C42H80NO8P
Molecular Weight (g/mol) 750
MDL Number MFCD00082428
InChI Key JLPULHDHAOZNQI-JLOPVYAASA-N
Synonym 3-sn-phosphatidylcholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholineShow MoreShow Less
PubChem CID 16213884
ChEBI CHEBI:86658
IUPAC Name [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC

Specifications

Acidity 35mg KOH/g max.
Color Tan or Yellow
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 1 kg
Merck Index 15, 5483
Solubility Information Solubility in water: negligible
Formula Weight 750
Percent Purity ≥97% (phospholipids) min. (as acetone insoluble)
Insoluble Matter 0.02% max. (in hexane)
Physical Form Granules
Chemical Name or Material L-α-Lecithin, Source: Soybean
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466455 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number413100010,413102500
Lot NumberA0466457 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number413100010,413102500
Lot NumberA0466454 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number413100010,413102500
Lot NumberA0466453 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number413100010,413102500
Lot NumberA0466450 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number413100010,413102500
Product Identifier
  • Lecithin, granular

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-alpha-Lecithin, granular, from soybean oil >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.