missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Aspartic acid, 98+% Product Code.: 11455623

Thermo Scientific Chemicals L-Aspartic acid, 98+%

Product Code. 11455623
5000 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11455623

Brand: Thermo Scientific Chemicals A13520.0I

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.

Solubility
Soluble in water(4.5g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-84-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00002616
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
Synonym l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acidShow MoreShow Less
PubChem CID 5960
ChEBI CHEBI:17053
SMILES NC(CC(O)=O)C(O)=O

Specifications

Melting Point >300°C (decomposition)
Beilstein 1723530
Merck Index 14,840
Solubility Information Soluble in water(4.5g/L).
Density 1.66
IUPAC Name (2S)-2-aminobutanedioic acid
Quantity 5000 g
Odor Odorless
Formula Weight 133.1
Optical Rotation +25° (c=2 in 5N HCl)
Percent Purity ≥98%
Chemical Name or Material L-Aspartic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461907 Certificate TypeCertificate of Analysis Date23/08/2024 Catalog NumberA13520.30,A13520.0I,A13520.0B
Lot NumberA0446963 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog NumberA13520.30,A13520.0I,A13520.0B
Lot NumberA0442008 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberA13520-22,A13520.30,A13520.0I,A13520.0B
Lot Number10239710 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13520-22,A13520.30,A13520.0I,A13520.0B
Lot Number10239840 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13520-22,A13520.30,A13520.0I,A13520.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Aspartic acid, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.