missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (10X)

Improve the reactivity of many antibodies in formalin-fixed tissues with the Epredia™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (10X).

Brand:  Epredia™ AP-9003-125

 View more versions of this product

Product Code. 12638286

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Citrate buffer, pH 6.0
  • 10X stock solution
  • Diluted 10-fold with distilled water before use
Storage Conditions:
  • Store at room temperature.
  • Stable for up to 18 months.
  • Contains no preservatives.
  • For periods longer than 3 months, store at 2-8°C.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Room temperature
Citrate Buffer
Immunohistochemistry
10X
125mL
6
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number74652467 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAP-9003-125
Lot Number74652467 Certificate TypeCertificate of Analysis Date23/05/2022 Catalog NumberAP-9003-125
Lot Number74184714 Certificate TypeCertificate of Analysis Date28/01/2022 Catalog NumberAP-9003-125
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (10X) > 125mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Reserach Use only