missing translation for 'onlineSavingsMsg'
Learn More
Cole-Parmer™ Essentials Collapsible Containers, LDPE Product Code.: 11779969
Add to basket

Cole-Parmer™ Essentials Collapsible Containers, LDPE

Product Code. 11779969 Shop All Cole-Parmer Products
10 L
Click to view available options
Capacity (Metric):
1 L
10 L
19 L
This item is not returnable. View return policy

Product Code. 11779969

Brand: Cole-Parmer™ Essentials 0610030

Add to basket
Add to basket
This item is not returnable. View return policy

LDPE Containers are collapsible when empty for easy storage.

  • These containers nest uniformly when empty and expand readily when filled with liquid.
  • They include polypropylene (PP) caps with injection-molded threads.
  • Replacement caps sold separately.

Specifications

Capacity (Metric) 10 L
Color Translucent
For Use With (Application) Containers nest uniformly when empty and expand readily when filled with liquid
Material LDPE
Screw Color White
Closure Color White
Closure Included Yes
Product Type Collapsible Container
Closure Material Polypropylene
Closure Type Screw Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Essentials Collapsible Containers, LDPE >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.