missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Le Mans S1P Shoe

Like its name suggests, you could wear the Le Mans for 24 hr. as the shoes offer optimal ventilation, perspiration and moisture removal with the Hydro-Tec™ antibacterial lining material.

£120.00 - £139.00

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Black, Gray, Orange
For Use With (Application) Logistics and Transport, Electronics, Manufacturing
Liner Material Hydro-Tec Antibacterial
View More Specs

Products 10
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17592476
View Documents
Emma Safety Footwear
MM70354739
39
£120.00
1 pair
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
17502486
View Documents
Emma Safety Footwear
MM70354740
40
£120.00
1 pair
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
17512486
View Documents
Emma Safety Footwear
MM70354741
41
£120.00
1 pair
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
17522486
View Documents
Emma Safety Footwear
MM70354742
42
£120.00
1 pair
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
17532486
View Documents
Emma Safety Footwear
MM70354743
43
£120.00
1 pair
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
17542486
View Documents
Emma Safety Footwear
MM70354744
44
£139.00
1 pair
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
17552486
View Documents
Emma Safety Footwear
MM70354745
45
£120.00
1 pair
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
17562486
View Documents
Emma Safety Footwear
MM70354746
46
£120.00
1 pair
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
17572486
View Documents
Emma Safety Footwear
MM70354747
47
£120.00
1 pair
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
17582486
View Documents
Emma Safety Footwear
MM70354748
48
£120.00
1 pair
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
Description

Description

  • Metal Free
  • Dry, fresh feet for up to 24 hr.The name already suggests it: you can safely wear LeMans for 24 hours.
  • The shoe offers optimal ventilation and moisture is immediately removed by the Hydro-Tec™ antibacterial lining.
  • LeMans is ideal for indoor working environments and work in transport and logistics.
  • LeMans looks sporty, that's how the shoe feels; soles with energy return and a fit that hugs the feet perfectly.
  • Industries:
    • Logistics & Transport
    • Electronics
    • Light Industry
  • Safety shoe
    • S1P-Safety shoe for dry conditions
Specifications

Specifications

ISO 20345:2011
Black, Gray, Orange
Hydro-Tec Antibacterial
Low-cut
Laces
Logistics and Transport, Electronics, Manufacturing
Standard Width
Safety Shoe
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Emma Safety Footwear Le Mans S1P Shoe

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.