missing translation for 'onlineSavingsMsg'
Learn More
Lecithin, 90%, soybean, Thermo Scientific Chemicals Product Code.: 15444229

Lecithin, 90%, soybean, Thermo Scientific Chemicals

Product Code. 15444229
250 g
Click to view available options
Quantity:
50 g
100 g
250 g
This item is not returnable. View return policy

Product Code. 15444229

Brand: Thermo Scientific Alfa Aesar J61675.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lecithin is used as a dietary supplement, food ingredient, and food additive. It is also used as a liquid carrier for plastics additives, dyes or pigments. Its primary functions are as an emulsifier, antioxidant, and release agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lecithin is used as a dietary supplement, food ingredient, and food additive. It is also used as a liquid carrier for plastics additives, dyes or pigments. Its primary functions are as an emulsifier, antioxidant, and release agent.

Solubility
Partly soluble in water, partly soluble in acetone, DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8002-43-5
Molecular Formula C42H80NO8P
Molecular Weight (g/mol) 758.075
MDL Number MFCD00147406
InChI Key JLPULHDHAOZNQI-JLOPVYAASA-N
Synonym 3-sn-phosphatidylcholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholineShow MoreShow Less
PubChem CID 16213884
ChEBI CHEBI:86658
IUPAC Name [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC

Specifications

Color Brown to Yellow
Odor Odorless
Quantity 250 g
Beilstein 5209585
Merck Index 14,5428
Solubility Information Partly soluble in water,partly soluble in acetone,DMSO.
Percent Purity 90%
Physical Form Solid
Chemical Name or Material Lecithin, soybean

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN21L034 Certificate TypeCertificate of Analysis Date26/02/2025 Catalog NumberJ61675.22,J61675.30,J61675.18
Lot NumberZ18K012 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog NumberJ61675.22,J61675.30,J61675.18
Lot NumberW17K058 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog NumberJ61675.22,J61675.30,J61675.18
Lot NumberU20K038 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog NumberJ61675.22,J61675.30,J61675.18
Lot NumberS19K028 Certificate TypeCertificate of Analysis Date25/06/2024 Catalog NumberJ61675.22,J61675.30,J61675.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lecithin, 90%, soybean, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.