Learn More
Thermo Scientific™ M-PER™ Mammalian Protein Extraction Reagent
Isolate total protein from cultured mammalian cells with this highly efficient lysis and extraction reagent.
Brand: Thermo Scientific™ 78501
273.34 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Thermo Scientific™ M-PER Mammalian Protein Extraction Reagent is designed to provide highly efficient total protein extraction from cultured mammalian cells.
The complete cell lysis reagent is a nondenaturing detergent formulation that dissolves cell membranes and extracts total soluble cellular protein in only 5 minutes. M-PER Reagent requires little or no mechanical disruption and yields more protein than freeze/thaw cycles and sonication. This mammalian cell lysis reagent is so efficient that adherent cells do not need to be scraped from the culture dish, enabling easy and direct lysis and analysis of cells grown in 24-well and 96-well plates. Resulting cell lysates from either adherent or suspension cells are compatible with many downstream assays including immunoassays, enzyme assays and a variety of common reporter assays.
Highlights:
Gentle – mild detergent lysis, yielding extracts that are immediately compatible with Coomassie™ (Bradford) and BCA protein assays or SDS-PAGE
Compatible – extracts soluble proteins in nondenatured state, enabling direct use in immunoprecipitation and other affinity purification procedures
Amine-free and dialyzable – formulation ensures compatibility with subsequent assay systems
Convenient – lyse adherent cells directly in plate or after scraping and washing in suspension
Non-denaturing – maintain luciferase, β-galactosidase, CAT and other reporter gene activities as well or better than other suppliers' products and freeze/thaw methods

Specifications
250 mL | |
Protein Extraction Reagent | |
Upon receipt store product at room temperature. |
Liquid | |
250 mL | |
M-PER™ |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.