missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ M-PER™ Mammalian Protein Extraction Reagent

Isolate total protein from cultured mammalian cells with this highly efficient lysis and extraction reagent.

Brand:  Thermo Scientific™ 78505

1094.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11884111

  • £1248.00 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

Thermo Scientific™ M-PER Mammalian Protein Extraction Reagent is designed to provide highly efficient total protein extraction from cultured mammalian cells.

The complete cell lysis reagent is a nondenaturing detergent formulation that dissolves cell membranes and extracts total soluble cellular protein in only 5 minutes. M-PER Reagent requires little or no mechanical disruption and yields more protein than freeze/thaw cycles and sonication. This mammalian cell lysis reagent is so efficient that adherent cells do not need to be scraped from the culture dish, enabling easy and direct lysis and analysis of cells grown in 24-well and 96-well plates. Resulting cell lysates from either adherent or suspension cells are compatible with many downstream assays including immunoassays, enzyme assays and a variety of common reporter assays.

Highlights:

  • Gentle – mild detergent lysis, yielding extracts that are immediately compatible with Coomassie™ (Bradford) and BCA protein assays or SDS-PAGE

  • Compatible – extracts soluble proteins in nondenatured state, enabling direct use in immunoprecipitation and other affinity purification procedures

  • Amine-free and dialyzable – formulation ensures compatibility with subsequent assay systems

  • Convenient – lyse adherent cells directly in plate or after scraping and washing in suspension

  • Non-denaturing – maintain luciferase, β-galactosidase, CAT and other reporter gene activities as well or better than other suppliers' products and freeze/thaw methods

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 L
Protein Extraction Reagent
Upon receipt store product at room temperature.
Liquid
1 L
M-PER™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL403693 Certificate TypeCertificate of Analysis Date27/12/2024 Catalog Number78505
Lot NumberZJ403478 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number78505
Lot NumberZH401983 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number78505
Lot NumberZG398781 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number78505
Lot NumberZF397898 Certificate TypeCertificate of Analysis Date06/07/2024 Catalog Number78505
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ M-PER™ Mammalian Protein Extraction Reagent > 1L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.