Learn More
Thermo Scientific™ MabCaptureC™ MiniChrom™ Pre-packed Column
Thermo Scientific™ MabCaptureC™ MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.
Brand: Thermo Scientific™ 5943662005
Description
MabCaptureC™ MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.
These pre-packed columns are ready to connect and have the following features:
- Packed with MabCaptureC™ Affinity Matrix, offering high dynamic binding capacity and high resolution over a wide range of flow rates
- Direct connection to standard chromatography systems
The 1 mL columns can be used for quick screenings of application feasibility and lab scale purification on a convenient and easy-to-use pre-packed column. The 10-cm bed height of the 5 mL column allows initial process development at a bench scale.
The pre-packed MabCaptureC™ Affinity Matrix contains engineered rProtein A that binds at the CH2-CH3 interface of the Fc-region of IgGs and shows cross binding to the VH3 domain of IgGs.
Features of MabCaptureC™ Affinity Matrix include:
- High dynamic mAb binding capacity of > 50 g/L at 4.8 min residence time
- Purification of mAbs and a subpopulation of scFv's and Fab fragments containing VH3 binding domains
- Clean and sanitize with standard sodium hydroxide agents for chromatography products
- Excellent scalability
- Non-animal derived
Main resin characteristics:
- Matrix: agarose-based, epoxide activated
- Average particle size: 75 ± 10 μm
- Ligand: recombinant C domain of Protein A
- Ligand coupling method: epoxide coupling
- Binding capacity: >55 g IgG/L resin at 5 min residence time and 10% breakthrough; >75 g IgG/L resin at 10 min. residence time and 10% breakthrough
- Elution conditions: 0.1 M glycine pH 3.0, or 20–100 mM sodium citrate or acetate, pH 3.0
- Cleaning conditions: 0.2 M sodium hydroxide for 15 minutes every cycle
- Column dimensions: 0.8 x 2 cm (1 mL), 0.8 x 10 cm (5 mL)
- Operating pressure: ≤ 3 bar
- Formulation buffer: 20% (v/v) ethanol
Specifications
5 mL | |
0.8 cm | |
10 cm | |
Agarose-based, Epoxide-activated | |
Agarose bead | |
5 mL |
>55 g IgG/L resin (5 min. residence time and 10% breakthrough), >75 g IgG/L resin (10 min. residence time and 10% breakthrough) | |
75 ±10 μm | |
Engineered rProtein A | |
≤3 bar | |
IgG mAbs, and a subpopulation of scFv and Fab containing VH3 domain |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.