missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Microbiome Ultra Nucleic Acid Isolation Kit, with bead tubes

The MagMAX Microbiome Ultra Nucleic Acid Isolation Kit enables efficient, high-throughput nucleic acid extraction from stool and soil.

Brand:  Applied Biosystems™ A42358

406.30 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 17180796

  • £483.00 / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

• 80 mL Lysis Buffer; room temperature
• 50 mL Binding Solution; room temperature
• 200 mL Wash Buffer; room temperature
• 20 mL Elution Solution; room temperature
• 4 mL Proteinase K; room temperature
• 2 mL Total Nucleic Acid Binding Beads; room temperature
• 100 Tubes; room temperature

Bacteria, Biofluids, Fecal, Soil, Swabs (Buccal or Skin), Yeast
RNA and DNA
Not High-throughput Compatible (Manual)
Room Temperature
Bacteria: ≤500 μL
Biofluids: ≤500 μL
Fecal: ≤100 mg (200 μL for samples in storage solution)
Soil: ≤250 mg
Yeast: ≤500 μL
Magnetic Bead
50 to 200 μL
RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization
100 preps
60 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ MagMAX™ Microbiome Ultra Nucleic Acid Isolation Kit, with bead tubes > 100 bead beating tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.