missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAX Efficiency™ Transformation Reagent for Algae
A buffer that enhances transformation efficiency for multiple strains of Chlamydomonas species
Brand: Invitrogen™ A24229
This item is not returnable.
View return policy
Description
MAX Efficiency™ Transformation Reagent for Algae is a buffer that enhances transformation efficiency for multiple strains of Chlamydomonas species. One of the biggest hurdles in research and development with Chlamydomonas has been the introduction of exogenous DNA into Chlamydomonas strains due to rigid cell walls. Methods such as glass bead agitation, electroporation, and microparticle bombardment are available but often result in low transformation efficiency.MAX Efficiency™ Transformation Reagent for Algae increases the permeability of the Chlamydomonas cell wall and facilitates increased delivery of DNA into the cell nucleus by electroporation. Simply grow the Chlamydomonas cells to early log phase, harvest by centrifugation, and wash twice with MAX Efficiency™ Transformation Reagent prior to electroporation. To date, we have seen >200-fold increase in transformation efficiency over previously recommended conditions in 10 different Chlamydomonas strains, including wild type and mutants, using circular or linear DNA, as well as PCR fragments.
- Obtain >1000 transformants/μg DNA for cell wall(+) strains
- Obtain >100 transformants/μg DNA for cell wall(-) strains
- Transform circular or linear DNA or PCR fragments
For Research Use Only. Not for use in diagnostic procedures.

Specifications
Chlamydomonas | |
Algae | |
Not High-throughput Compatible (Manual) | |
GeneArt™ | |
40 reactions |
Contains: 1 bottle Transformation Reagent, 250 mL |
|
Gel Electrophoresis | |
40 Reactions | |
Transformation Reagent | |
Hygromycin |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
GeneArt MAX Efficiency Transformation Reagent for Algae
Version: Rev A.0
File size: 133.4 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ MAX Efficiency™ Transformation Reagent for Algae > 40 reactions
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction