Learn More
Micronova™ MegaClean™ Heavy-Duty Cleaner
A non-flammable, heavy duty cleaner and degreaser that removes grease, adhesives and photo-resist stains from most surfaces. Available in 1 or 5 gallon bottles and a 1 quart spray bottle.
Brand:  Micronova™ MC1-Q
 View more versions of this product
Product Code. 13157736
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The Micronova MegaClean™ Heavy-Duty Cleaner offers an effective alternative to toxic, flammable, solvent-based cleaners, such as acetone and isopropyl alcohol. Filtered to 0.1 microns, this cleaner is ideal for semiconductor, electronics, aerospace and clean-build operations.
  • Non-flammable, heavy duty cleanroom degreaser
  • Removes grease, adhesives and photo-resist stains from most surfaces
  • Ideal for semiconductor, electronics, aerospace and clean-build operations
  • Filtered to 0.1 microns
  • Available in 1 or 5 gallon refill bottles and a 1 quart spray bottle
  • Biodegradable
  • Water soluble
  • Effective at low concentrations
Specifications
Specifications
Heavy Duty Cleaner
Heavy Duty Detergent, Degreaser
An effective alternative to toxic and flammable solvent based cleaners
1 qt.
Non-flammable, biodegradeable
0.1 μm
Mops and Buckets or Wipes and Sponge
0.9 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number038878-000_221109167 Certificate TypeCertificate of Compliance Date28/06/2023 Catalog NumberMC1-Q
Lot Number033512-000_220201398 Certificate TypeCertificate of Compliance Date21/07/2022 Catalog NumberMC1-Q
Lot Number024431-000_2009008505 Certificate TypeCertificate of Compliance Date19/03/2021 Catalog NumberMC1-Q
Special Offers
Special Offers
Product Content Correction
Product Title
Micronova™ MegaClean™ Heavy-Duty Cleaner > MegaClean Heavy Duty Cleanroom Detergent and degreaser, biodegradable, 1 qt