missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ MegaClean™ Heavy-Duty Cleaner
BIOPHARMA

A non-flammable, heavy duty cleaner and degreaser that removes grease, adhesives and photo-resist stains from most surfaces. Available in 1 or 5 gallon bottles and a 1 quart spray bottle.

Brand:  Micronova™ MC1-G


 View more versions of this product

Product Code. 11932475

  • £441.00 / 15.14 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Micronova MegaClean™ Heavy-Duty Cleaner offers an effective alternative to toxic, flammable, solvent-based cleaners, such as acetone and isopropyl alcohol. Filtered to 0.1 microns, this cleaner is ideal for semiconductor, electronics, aerospace and clean-build operations.

  • Non-flammable, heavy duty cleanroom degreaser
  • Removes grease, adhesives and photo-resist stains from most surfaces
  • Ideal for semiconductor, electronics, aerospace and clean-build operations
  • Filtered to 0.1 microns
  • Available in 1 or 5 gallon refill bottles and a 1 quart spray bottle
  • Biodegradable
  • Water soluble
  • Effective at low concentrations
Specifications

Specifications

Non-sterile
Heavy Soils, Adhesive Removal in and Around Cleanroom
1 gal.
Heavy Duty Detergent, Degreaser
Mops and Buckets or Wipes and Sponge
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : 2-(2-butoxyethoxy)ethanol, alcohols, C9-11, Ethoxylated

SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number042691-000 Certificate TypeCertificate of Compliance Date03/06/2024 Catalog NumberMC1-G
Lot Number041044-000 Certificate TypeCertificate of Compliance Date14/02/2024 Catalog NumberMC1-G
Lot Number031281-000_ 220201398 Certificate TypeCertificate of Compliance Date14/07/2022 Catalog NumberMC1-G
Lot Number027788-000 Certificate TypeCertificate of Compliance Date15/11/2021 Catalog NumberMC1-G
Lot Number035901-000_221109167 Certificate TypeDeclaration of Conformity Date01/02/2023 Catalog NumberMC1-G
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Micronova™ MegaClean™ Heavy-Duty Cleaner > MegaClean Heavy Duty Cleanroom Detergent and degreaser, biodegradable, 1 gal.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.